Product Details

SNP ID
rs7980833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80796647 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCTGAGAGTGCCAAAAATATCT[C/G]GTTTATATTTGGGGAAATTGACCTA
Phenotype
MIM: 603380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIN7A PubMed Links

Gene Details

Gene
LIN7A
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324423.1 1750 UTR 3 NP_001311352.1
NM_004664.3 1750 UTR 3 NP_004655.1
XM_011538928.2 1750 UTR 3 XP_011537230.1

View Full Product Details