Product Details

SNP ID
rs6748431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208170934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATCAGTTTGGTCCAATTTACAAT[C/T]TCACACCTACTTTGTGCTTTGGCTG
Phenotype
MIM: 615536 MIM: 123660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf80 PubMed Links

Gene Details

Gene
C2orf80
Gene Name
chromosome 2 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099334.2 Intron NP_001092804.1
XM_017004074.1 Intron XP_016859563.1
XM_017004075.1 Intron XP_016859564.1
XM_017004076.1 Intron XP_016859565.1
XM_017004077.1 Intron XP_016859566.1
XM_017004078.1 Intron XP_016859567.1
Gene
CRYGA
Gene Name
crystallin gamma A
There are no transcripts associated with this gene.

View Full Product Details