Product Details

SNP ID
rs7696066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145140562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAATTTTGGAAACTAAAAAGGA[A/G]GGAGGTGTAAATTCCTTAAAAATAA
Phenotype
MIM: 611744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD4 PubMed Links

Gene Details

Gene
OTUD4
Gene Name
OTU deubiquitinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102653.1 Intron NP_001096123.1
NM_017493.6 Intron NP_059963.1
XM_005263079.4 Intron XP_005263136.1
XM_011532040.2 Intron XP_011530342.2
XM_011532041.2 Intron XP_011530343.2
XM_011532042.2 Intron XP_011530344.1

View Full Product Details