Product Details

SNP ID
rs7172816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38698568 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATGTCAAGGCCTAGTTACAGGGA[C/T]GTCAAACTAGGAGGTTGATGTCGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf53 PubMed Links

Gene Details

Gene
C15orf53
Gene Name
chromosome 15 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207444.2 573 UTR 3 NP_997327.1

View Full Product Details