Product Details

SNP ID
rs7872832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:105448898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGTGGAATGTTTTATACTTTCCG[A/G]ACAGTTGAGGAAAGGTTATATAATC
Phenotype
MIM: 609829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSD1L PubMed Links

Gene Details

Gene
FSD1L
Gene Name
fibronectin type III and SPRY domain containing 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145313.2 Intron NP_001138785.1
NM_001287191.1 Intron NP_001274120.1
NM_001287192.1 Intron NP_001274121.1
NM_031919.4 Intron NP_114125.1
XM_005252254.2 Intron XP_005252311.1
XM_005252257.3 Intron XP_005252314.1
XM_005252260.3 Intron XP_005252317.1
XM_011519077.2 Intron XP_011517379.1
XM_011519078.2 Intron XP_011517380.2
XM_011519079.2 Intron XP_011517381.2
XM_011519080.2 Intron XP_011517382.1
XM_011519081.2 Intron XP_011517383.1
XM_017015182.1 Intron XP_016870671.1
XM_017015183.1 Intron XP_016870672.1
XM_017015184.1 Intron XP_016870673.1
XM_017015185.1 Intron XP_016870674.1
XM_017015186.1 Intron XP_016870675.1
XM_017015187.1 Intron XP_016870676.1

View Full Product Details