Product Details

SNP ID
rs7333241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51862822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTAGGCAAAGAGCCATACAACA[C/T]AGAAAATCAAATTTCCAAAATGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC70 PubMed Links

Gene Details

Gene
CCDC70
Gene Name
coiled-coil domain containing 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031290.2 Intron NP_112580.2

View Full Product Details