Product Details
- SNP ID
-
rs6056237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:960752 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCTCGGCCAGGCCTGCAGGTTCA[A/C]CTGGGGCCCCCCAATCAGCATGTCG
- Phenotype
-
MIM: 610573
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RSPO4
PubMed Links
Gene Details
- Gene
- RSPO4
- Gene Name
- R-spondin 4
View Full Product Details