Product Details
- SNP ID
-
rs7711132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:78487839 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCAGTCTGCTTTCAGGATCAACAC[C/T]AGTAACTACAAAAACTTAACTCCCA
- Phenotype
-
MIM: 609718
MIM: 606911
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LHFPL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76221473] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LHFPL2
- Gene Name
- lipoma HMGIC fusion partner-like 2
- Gene
- SCAMP1
- Gene Name
- secretory carrier membrane protein 1
There are no transcripts associated with this gene.
View Full Product Details