Product Details
- SNP ID
-
rs6768709
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:134490024 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTGGTAAATAATTTTCCTGGCTC[A/G]TTTATAGATTTCTGGTCTTCAAGGA
- Phenotype
-
MIM: 614484
MIM: 614724
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANAPC13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77039188] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANAPC13
- Gene Name
- anaphase promoting complex subunit 13
There are no transcripts associated with this gene.
- Gene
- CEP63
- Gene Name
- centrosomal protein 63
View Full Product Details