Product Details

SNP ID
rs6854175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44702418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATTAGGGACATGAACCCAAGTC[A/G]TTAAATAAAAATAAGATATTTGTAA
Phenotype
MIM: 613222
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNPDA2 PubMed Links

Gene Details

Gene
GNPDA2
Gene Name
glucosamine-6-phosphate deaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270880.1 1588 UTR 3 NP_001257809.1
NM_001270881.1 1588 UTR 3 NP_001257810.1
NM_138335.2 1588 UTR 3 NP_612208.1
Gene
GUF1
Gene Name
GUF1 homolog, GTPase
There are no transcripts associated with this gene.

View Full Product Details