Product Details

SNP ID
rs7887769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:7158107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGAAATAAAAACAAATGACATG[A/G]CATGCATCTCTCTTAGCCTCAGTCC
Phenotype
MIM: 306480 MIM: 300747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PUDP PubMed Links

Gene Details

Gene
PUDP
Gene Name
pseudouridine 5'-phosphatase
There are no transcripts associated with this gene.

Gene
STS
Gene Name
steroid sulfatase (microsomal), isozyme S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000351.5 Intron NP_000342.2
NM_001320750.1 Intron NP_001307679.1
NM_001320751.1 Intron NP_001307680.1
NM_001320752.1 Intron NP_001307681.1
NM_001320753.1 Intron NP_001307682.1
NM_001320754.1 Intron NP_001307683.1

View Full Product Details