Product Details

SNP ID
rs7717213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179919466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAGATCATTCTGTCTTTCACAA[C/T]TGCTGGCCATCCCCCTGAAATCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF130 PubMed Links

Gene Details

Gene
RNF130
Gene Name
ring finger protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280801.1 2539 Intron NP_001267730.1
NM_018434.5 2539 Intron NP_060904.2
XM_011534593.2 2539 UTR 3 XP_011532895.1

View Full Product Details