Product Details

SNP ID
rs7479477
Assay Type
Functionally Tested
NCBI dbSNP Submissions
27
Location
Chr.11:5351881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTATGACTGTTTCATTACCATCC[A/G]CAGCCCCTTAAGATATACCTCTATC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 374 Intron NP_001005567.2
Gene
OR51B6
Gene Name
olfactory receptor family 51 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004750.1 374 Missense Mutation CAC,CGC H125R NP_001004750.1

View Full Product Details