Product Details
- SNP ID
-
rs7405662
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41630613 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGCCAGCACGAAGCTGGAATTACC[A/G]ACCACAGCCACCAGGATCTGGGAGA
- Phenotype
-
MIM: 148069
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT17
PubMed Links
Gene Details
- Gene
- KRT17
- Gene Name
- keratin 17
There are no transcripts associated with this gene.
- Gene
- KRT42P
- Gene Name
- keratin 42 pseudogene
There are no transcripts associated with this gene.
View Full Product Details