Product Details

SNP ID
rs7885989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101836705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACACTGACCCCAAGCTGGGATG[C/G]GTGCTCCCACCCACAACATAGCACT
Phenotype
MIM: 300319
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NXF5 PubMed Links

Gene Details

Gene
NXF5
Gene Name
nuclear RNA export factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032946.2 Intron NP_116564.2

View Full Product Details