Product Details

SNP ID
rs8078382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19377947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGAAGTCAGTGGAGGGTTCGGC[C/T]GGACGCTCTAGAATCCCGGAGGACC
Phenotype
MIM: 614144 MIM: 602521 MIM: 600596
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B9D1 PubMed Links

Gene Details

Gene
B9D1
Gene Name
B9 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243473.2 227 Intron NP_001230402.1
NM_001243475.2 227 UTR 5 NP_001230404.1
NM_001321214.1 227 Intron NP_001308143.1
NM_001321215.1 227 Intron NP_001308144.1
NM_001321216.1 227 Intron NP_001308145.1
NM_001321217.1 227 Intron NP_001308146.1
NM_001321218.1 227 Intron NP_001308147.1
NM_001321219.1 227 Intron NP_001308148.1
NM_015681.4 227 Intron NP_056496.1
XM_005256607.2 227 Intron XP_005256664.1
XM_005256610.1 227 Intron XP_005256667.1
XM_017024452.1 227 UTR 5 XP_016879941.1
Gene
MAPK7
Gene Name
mitogen-activated protein kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002749.3 227 Intron NP_002740.2
NM_139032.2 227 UTR 5 NP_620601.1
NM_139033.2 227 UTR 5 NP_620602.2
NM_139034.2 227 Intron NP_620603.2
XM_006721557.3 227 Intron XP_006721620.1
XM_006721558.3 227 UTR 5 XP_006721621.1
XM_006721559.3 227 Intron XP_006721622.1
XM_011523957.2 227 Intron XP_011522259.1
Gene
MFAP4
Gene Name
microfibrillar associated protein 4
There are no transcripts associated with this gene.

View Full Product Details