Product Details

SNP ID
hCV29058904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26442414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAACTTCCGATCTTCAGTTTAGC[A/T]TGCATCTTTTTAGACTTTTTCAGGC
Phenotype
MIM: 613594 MIM: 613595
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BTN3A2 PubMed Links

Gene Details

Gene
BTN3A2
Gene Name
butyrophilin subfamily 3 member A2
There are no transcripts associated with this gene.

Gene
BTN3A3
Gene Name
butyrophilin subfamily 3 member A3
There are no transcripts associated with this gene.

View Full Product Details