Product Details

SNP ID
rs8071908
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39258854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGAATATTATTAACCACATCT[A/G]GAAGTATGTGTGTATGCACGTATGT
Phenotype
MIM: 609086
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FBXL20 PubMed Links

Gene Details

Gene
FBXL20
Gene Name
F-box and leucine rich repeat protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184906.1 4171 UTR 3 NP_001171835.1
NM_032875.2 4171 UTR 3 NP_116264.2
XM_005257746.3 4171 UTR 3 XP_005257803.2
XM_005257747.3 4171 UTR 3 XP_005257804.2

View Full Product Details