Product Details
- SNP ID
-
rs4646241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:18389186 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAATCTGGATTGCAACATTTTAAT[C/T]CCAGGTGTCAGGTTTCCAACAATCA
- Phenotype
-
MIM: 612182
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NAT2
PubMed Links
Gene Details
- Gene
- NAT2
- Gene Name
- N-acetyltransferase 2
View Full Product Details