Product Details

SNP ID
hCV29072030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216415273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGCCTCCCAGGGATGCTAAGT[G/T]AGAGGCCAAGACAGCAAAAGCCTCC
Phenotype
MIM: 606622
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SMARCAL1 PubMed Links

Gene Details

Gene
SMARCAL1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127207.1 713 Nonsense Mutation TGA,TTA *190L NP_001120679.1
NM_014140.3 713 Nonsense Mutation TGA,TTA *190L NP_054859.2

View Full Product Details