Product Details

SNP ID
rs4682534
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114315251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAAGATTGGAGGGTATAACACTG[C/T]AGTCATTCAGCCCAGGTTCCTCTTA
Phenotype
MIM: 612859 MIM: 606025
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR568 PubMed Links

Gene Details

Gene
MIR568
Gene Name
microRNA 568
There are no transcripts associated with this gene.

Gene
TIGIT
Gene Name
T-cell immunoreceptor with Ig and ITIM domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173799.3 26201 Intron NP_776160.2
Gene
ZBTB20
Gene Name
zinc finger and BTB domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164342.2 26201 UTR 3 NP_001157814.1
NM_001164343.2 26201 UTR 3 NP_001157815.1
NM_001164344.2 26201 UTR 3 NP_001157816.1
NM_001164345.2 26201 UTR 3 NP_001157817.1
NM_001164346.2 26201 UTR 3 NP_001157818.1
NM_001164347.2 26201 UTR 3 NP_001157819.1
NM_015642.5 26201 UTR 3 NP_056457.3

View Full Product Details