Product Details

SNP ID
rs8006919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96377912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCCAGTACATGTTGCTATGAT[G/T]ATGCTGGAATTAAAATACAAAGCAT
Phenotype
MIM: 616605
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GSKIP PubMed Links

Gene Details

Gene
GSKIP
Gene Name
GSK3B interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271904.1 Intron NP_001258833.1
NM_001271905.1 Intron NP_001258834.1
NM_001271906.1 Intron NP_001258835.1
NM_016472.4 Intron NP_057556.2
XM_017021365.1 Intron XP_016876854.1

View Full Product Details