Product Details

SNP ID
rs7478515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12144412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTACTTTGTTTACTTAATATCT[A/T]ACATGAGCTATTAATGGCCATGTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC61A2 PubMed Links

Gene Details

Gene
SEC61A2
Gene Name
Sec61 translocon alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142627.2 Intron NP_001136099.1
NM_001142628.1 Intron NP_001136100.1
NM_018144.3 Intron NP_060614.2

View Full Product Details