Product Details
- SNP ID
-
rs6833237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:15974982 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGTCGGCTAATATCCAGAGCCCAG[C/T]TGGTGTCATCAATGGAAGCTAAGTG
- Phenotype
-
MIM: 604365
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROM1
PubMed Links
Gene Details
- Gene
- PROM1
- Gene Name
- prominin 1
View Full Product Details