Product Details

SNP ID
rs7579175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31175546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCTATTCTCCTAGACCTGAGCC[A/G]GCCACTGTGGAAGGATTGCCTGGGA
Phenotype
MIM: 610229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145122.1 Intron NP_001138594.1
NM_001321270.1 Intron NP_001308199.1
XM_011532863.2 Intron XP_011531165.1
XM_011532864.2 Intron XP_011531166.1
XM_011532865.1 Intron XP_011531167.1
XM_011532866.2 Intron XP_011531168.1

View Full Product Details