Product Details

SNP ID
rs8029808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101843711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAATTATTCTGCTATGATAGTTC[A/G]TTGGACGTTTAGTTGTTTTTTTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4F13P PubMed Links

Gene Details

Gene
OR4F13P
Gene Name
olfactory receptor family 4 subfamily F member 13 pseudogene
There are no transcripts associated with this gene.

View Full Product Details