Product Details

SNP ID
rs6535428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83035547 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTGTTCAGTTCTTGATAGAATC[C/T]TTCTGGTCCCTTAGGATTTTGGAAC
Phenotype
MIM: 616008
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPS4 PubMed Links

Gene Details

Gene
COPS4
Gene Name
COP9 signalosome subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258006.1 Intron NP_001244935.1
NM_016129.2 Intron NP_057213.2
XM_011532019.1 Intron XP_011530321.1

View Full Product Details