Product Details

SNP ID
rs7450789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111495772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGGAAGGTTATACCGTGTTATG[G/T]ATTAATCTCTACCTTATGCCAAAGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRAF3IP2-AS1 PubMed Links

Gene Details

Gene
TRAF3IP2-AS1
Gene Name
TRAF3IP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details