Product Details
- SNP ID
-
rs7590874
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:76750644 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCTTATGGCTCCCTATACTGCAGG[G/T]AGTTAACCAGTCTTAAATATAATTC
- Phenotype
-
MIM: 610870
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRTM4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76543253,rs77073950] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRTM4
- Gene Name
- leucine rich repeat transmembrane neuronal 4
View Full Product Details