Product Details

SNP ID
rs7969185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101162948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAAAGTGCTGTAAGGAAAGTAAA[A/G]CAAGATGATGTGCTATTAACAGGGT
Phenotype
MIM: 608044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC5A8 PubMed Links

Gene Details

Gene
SLC5A8
Gene Name
solute carrier family 5 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145913.3 Intron NP_666018.3
XM_017018910.1 Intron XP_016874399.1

View Full Product Details