Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193528.1 | Intron | NP_001180457.1 | |||
NM_024583.4 | Intron | NP_078859.2 | |||
XM_005246853.2 | Intron | XP_005246910.1 | |||
XM_005246854.4 | Intron | XP_005246911.1 | |||
XM_005246855.2 | Intron | XP_005246912.1 | |||
XM_005246856.2 | Intron | XP_005246913.1 | |||
XM_005246857.2 | Intron | XP_005246914.1 | |||
XM_011511839.2 | Intron | XP_011510141.1 | |||
XM_017004909.1 | Intron | XP_016860398.1 | |||
XM_017004910.1 | Intron | XP_016860399.1 |