Product Details
- SNP ID
-
rs7161062
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:100377931 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCACCTGTTGGGGGATGGAGGCG[A/G]AGCTTGGCGTGTTTGATTAAGGTTG
- Phenotype
-
MIM: 191050
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WARS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7161066] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WARS
- Gene Name
- tryptophanyl-tRNA synthetase
There are no transcripts associated with this gene.
- Gene
- WDR25
- Gene Name
- WD repeat domain 25
View Full Product Details