Product Details

SNP ID
rs6578927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8093111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTTAAACAGATGGTCACGGGG[G/T]TGCTTTGTGGAAGGAAGCCAAATTC
Phenotype
MIM: 610509 MIM: 601197
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RIC3 PubMed Links

Gene Details

Gene
RIC3
Gene Name
RIC3 acetylcholine receptor chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135109.2 Intron NP_001128581.1
NM_001206671.2 Intron NP_001193600.1
NM_001206672.2 Intron NP_001193601.1
NM_024557.4 Intron NP_078833.3
XM_006718317.3 Intron XP_006718380.1
XM_006718318.3 Intron XP_006718381.1
XM_006718321.3 Intron XP_006718384.1
XM_017018287.1 Intron XP_016873776.1
XM_017018288.1 Intron XP_016873777.1
XM_017018289.1 Intron XP_016873778.1
XM_017018290.1 Intron XP_016873779.1
Gene
TUB
Gene Name
tubby bipartite transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003320.4 Intron NP_003311.2
NM_177972.2 Intron NP_813977.1
XM_005253109.3 Intron XP_005253166.1
XM_011520344.2 Intron XP_011518646.1

View Full Product Details