Product Details

SNP ID
hCV29140107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:109869700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTCTAGATCCCAGTTCATTAA[A/G]GATTTTCTTTCTGGCTTTGCAGGGA
Phenotype
MIM: 602443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPNPEP1 PubMed Links

Gene Details

Gene
XPNPEP1
Gene Name
X-prolyl aminopeptidase 1
There are no transcripts associated with this gene.

View Full Product Details