Product Details

SNP ID
rs7802626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:14147578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAATCACTAAATGCTATTTGTTA[C/T]TCAAAGAAAGATGAATGAATCAATA
Phenotype
MIM: 604070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKB PubMed Links

Gene Details

Gene
DGKB
Gene Name
diacylglycerol kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004080.2 4155 UTR 3 NP_004071.1
NM_145695.2 4155 Intron NP_663733.1
XM_005249628.2 4155 Intron XP_005249685.1
XM_005249629.2 4155 Intron XP_005249686.1
XM_005249630.2 4155 Intron XP_005249687.1
XM_005249631.3 4155 Intron XP_005249688.1
XM_011515153.2 4155 Intron XP_011513455.1
XM_011515154.2 4155 Intron XP_011513456.1
XM_011515156.2 4155 Intron XP_011513458.1
XM_011515157.2 4155 Intron XP_011513459.1
XM_011515158.2 4155 Intron XP_011513460.1
XM_017011783.1 4155 Intron XP_016867272.1
XM_017011784.1 4155 Intron XP_016867273.1
XM_017011785.1 4155 Intron XP_016867274.1
XM_017011786.1 4155 Intron XP_016867275.1
XM_017011787.1 4155 Intron XP_016867276.1
XM_017011788.1 4155 Intron XP_016867277.1
XM_017011789.1 4155 Intron XP_016867278.1
XM_017011790.1 4155 Intron XP_016867279.1
XM_017011791.1 4155 Intron XP_016867280.1
XM_017011792.1 4155 Intron XP_016867281.1

View Full Product Details