Product Details
- SNP ID
-
rs6951212
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:17305374 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGCACTGAAGCTGGGGGAAGGAAA[C/T]GAAGAATGAATGGGGTTAAGAGTTG
- Phenotype
-
MIM: 600253
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AHR
PubMed Links
Gene Details
- Gene
- AHR
- Gene Name
- aryl hydrocarbon receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001621.4 |
|
Intron |
|
|
NP_001612.1 |
View Full Product Details