Product Details

SNP ID
rs7989972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:43214041 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACAGCTTCCATCTTTTTTCCAG[A/C]GTGGCTCCCACACCCGTGAATTCCT
Phenotype
MIM: 610914
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENOX1 PubMed Links

Gene Details

Gene
ENOX1
Gene Name
ecto-NOX disulfide-thiol exchanger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127615.1 2494 Silent Mutation NP_001121087.1
NM_001242863.1 2494 Silent Mutation NP_001229792.1
NM_017993.3 2494 Silent Mutation NP_060463.2
XM_005266439.3 2494 Silent Mutation XP_005266496.1
XM_011535126.2 2494 Silent Mutation XP_011533428.1
XM_011535127.2 2494 Silent Mutation XP_011533429.1
XM_011535128.2 2494 Silent Mutation XP_011533430.1
XM_011535132.2 2494 Silent Mutation XP_011533434.1
XM_017020637.1 2494 Silent Mutation XP_016876126.1
XM_017020638.1 2494 Silent Mutation XP_016876127.1
XM_017020639.1 2494 Silent Mutation XP_016876128.1
XM_017020640.1 2494 Silent Mutation XP_016876129.1
XM_017020641.1 2494 Intron XP_016876130.1
XM_017020642.1 2494 Intron XP_016876131.1

View Full Product Details