Product Details

SNP ID
rs7934948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78436328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCACAAATTACACAGGGAGTGA[C/T]TTAATGATTATAGACAACGTAAAAT
Phenotype
MIM: 612803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928865 PubMed Links

Gene Details

Gene
LOC101928865
Gene Name
uncharacterized LOC101928865
There are no transcripts associated with this gene.

Gene
NARS2
Gene Name
asparaginyl-tRNA synthetase 2, mitochondrial (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243251.1 2118 UTR 3 NP_001230180.1
NM_024678.5 2118 UTR 3 NP_078954.4
XM_011545253.1 2118 UTR 3 XP_011543555.1
XM_017018302.1 2118 Intron XP_016873791.1
XM_017018303.1 2118 UTR 3 XP_016873792.1
XM_017018304.1 2118 UTR 3 XP_016873793.1

View Full Product Details