Product Details

SNP ID
rs7150536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50877047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCATGGACAAATCGTCACTTGT[A/G]TGCTGACAGCTCCTGATTGGGTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD12B PubMed Links

Gene Details

Gene
ABHD12B
Gene Name
abhydrolase domain containing 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206673.1 Intron NP_001193602.1
NM_181533.3 Intron NP_853511.2
NM_181814.1 Intron NP_861535.1
XM_006720052.1 Intron XP_006720115.1
XM_011536472.1 Intron XP_011534774.1
XM_011536474.2 Intron XP_011534776.1
XM_017021031.1 Intron XP_016876520.1

View Full Product Details