Product Details

SNP ID
rs8020560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49787005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTAGTTCTCTGAAGTAGAGGAG[A/G]TGGGAACTGAAACTTTCATATAAAG
Phenotype
MIM: 611280 MIM: 608378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC2 PubMed Links

Gene Details

Gene
KLHDC2
Gene Name
kelch domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014315.2 Intron NP_055130.1
Gene
NEMF
Gene Name
nuclear export mediator factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011537317.2 Intron XP_011535619.1
XM_011537318.2 Intron XP_011535620.1
XM_017021760.1 Intron XP_016877249.1
XM_017021761.1 Intron XP_016877250.1

View Full Product Details