Product Details

SNP ID
rs7831370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42401557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAAATCATGAAGTATCCTATCA[C/T]TTGAACTTATAATGAGTAATAGATG
Phenotype
MIM: 610029
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VDAC3 PubMed Links

Gene Details

Gene
VDAC3
Gene Name
voltage dependent anion channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135694.2 Intron NP_001129166.1
NM_005662.6 Intron NP_005653.3
XM_006716394.1 Intron XP_006716457.1

View Full Product Details