Product Details

SNP ID
rs7741430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25849677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCCTCAACATACCTGAAAAAAA[C/T]GGCCACAGGTCTATCTGTGGTTCTT
Phenotype
MIM: 611034
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC17A3 PubMed Links

Gene Details

Gene
SLC17A3
Gene Name
solute carrier family 17 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098486.1 Intron NP_001091956.1
NM_006632.3 Intron NP_006623.2

View Full Product Details