Product Details

SNP ID
rs7957702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14505744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGACTCATGTTTAACATGGAAT[C/T]GCACTGCTTGAATAATATGTTCTTT
Phenotype
MIM: 613644
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF7IP PubMed Links

Gene Details

Gene
ATF7IP
Gene Name
activating transcription factor 7 interacting protein
There are no transcripts associated with this gene.

Gene
PLBD1
Gene Name
phospholipase B domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024829.5 Intron NP_079105.4

View Full Product Details