Product Details
- SNP ID
-
rs7231013
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:50976485 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATATGCAGGGGAACCACCCTTTAT[C/T]AACCCGTCAGATCTCATGAGACTTA
- Phenotype
-
MIM: 608079
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELAC1
PubMed Links
Gene Details
- Gene
- ELAC1
- Gene Name
- elaC ribonuclease Z 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018696.2 |
|
Intron |
|
|
NP_061166.1 |
View Full Product Details