Product Details

SNP ID
rs7183006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89249093 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTTGTTTTACTCCATCAAGATC[A/C]TTTTGTATGATTCTGCTGACCGTAT
Phenotype
MIM: 611360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FANCI PubMed Links

Gene Details

Gene
FANCI
Gene Name
Fanconi anemia complementation group I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113378.1 Intron NP_001106849.1
NM_018193.2 Intron NP_060663.2
XM_011521756.2 Intron XP_011520058.1
XM_011521757.2 Intron XP_011520059.1
XM_011521764.2 Intron XP_011520066.1
XM_011521767.2 Intron XP_011520069.1

View Full Product Details