Product Details

SNP ID
rs7255420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12047024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGGTGAGTCCAGGGAGAAAGTC[A/G]TGCTGCCTGGACTGCCCTTAATGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928464 PubMed Links

Gene Details

Gene
LOC101928464
Gene Name
uncharacterized LOC101928464
There are no transcripts associated with this gene.

Gene
ZNF878
Gene Name
zinc finger protein 878
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080404.2 Intron NP_001073873.2
XM_017027195.1 Intron XP_016882684.1
XM_017027196.1 Intron XP_016882685.1

View Full Product Details