Product Details
- SNP ID
-
rs7255351
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12074785 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGATCACTTGAGTCCAGGAGTTC[A/G]AGGGTGCTGTGAACCATGATGATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF844
PubMed Links
Gene Details
- Gene
- ZNF844
- Gene Name
- zinc finger protein 844
View Full Product Details