Product Details

SNP ID
rs7878811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71106665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGGCCTATAGGGTTGGTGGACT[A/G]GAGAGGCAGATGGATTTCATAGGCT
Phenotype
MIM: 300033 MIM: 308380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025265.2 124 UTR 5 NP_001020436.1
XM_005262244.3 124 UTR 5 XP_005262301.1
Gene
FOXO4
Gene Name
forkhead box O4
There are no transcripts associated with this gene.

Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000206.2 124 Intron NP_000197.1

View Full Product Details