Product Details

SNP ID
rs8112935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56438598 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGATTCGGCCTATTAGGATATAA[C/T]TGAGTTCCAAGCAGTTGGGAGGACA
Phenotype
MIM: 611024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF667 PubMed Links

Gene Details

Gene
ZNF667
Gene Name
zinc finger protein 667
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321355.1 6832 Intron NP_001308284.1
NM_001321356.1 6832 Intron NP_001308285.1
NM_022103.3 6832 Intron NP_071386.3
XM_011527208.1 6832 UTR 3 XP_011525510.1
XM_011527209.1 6832 UTR 3 XP_011525511.1

View Full Product Details