Product Details

SNP ID
rs6573950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70327177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAAAAAAAAAGCTGTGAAAACA[C/T]AGGATAGCTTATATCCAGAGACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX16 PubMed Links

Gene Details

Gene
COX16
Gene Name
COX16, cytochrome c oxidase assembly homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204090.1 Intron NP_001191019.1
NM_016468.6 Intron NP_057552.1
Gene
SYNJ2BP-COX16
Gene Name
SYNJ2BP-COX16 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202547.2 Intron NP_001189476.1
NM_001202548.2 Intron NP_001189477.1
NM_001202549.2 Intron NP_001189478.1

View Full Product Details